Learn More
MSC Huot Drill Stand, 1/2 to 1 in. Capacity, Jobbers/Stub/Machine Screw Drills
Catalog No. 5000534263
Encompass
This drill stand is designed to hold jobber, stub, and machine screw length drills. It accommodates drills from 1/2 to 1 in. by 64ths increments. The stand features permanently embossed fractional sizing for quick selection and a durable powder coat finish.
  • Holds 1/2 to 1 in. drills in 1/64 in. increments.
  • Compatible with jobber, stub, and screw machine length drills.
  • Designed to hold drills by the flute.
  • Features permanently embossed fractional sizing for easy drill selection.
  • Durable powder coat finish.
  • Steel construction with ecoloclad plastic finish for long, usable life.
  • Suitable for fractional inch sizes.
  • Manufactured in the United States.
Catalog No. 50-005-34263 Supplier MSC Supplier No. 00922468
This drill stand is designed to hold jobber, stub, and machine screw length drills. It accommodates drills from 1/2 to 1 in. by 64ths increments. The stand features permanently embossed fractional sizing for quick selection and a durable powder coat finish.
  • Holds 1/2 to 1 in. drills in 1/64 in. increments.
  • Compatible with jobber, stub, and screw machine length drills.
  • Designed to hold drills by the flute.
  • Features permanently embossed fractional sizing for easy drill selection.
  • Durable powder coat finish.
  • Steel construction with ecoloclad plastic finish for long, usable life.
  • Suitable for fractional inch sizes.
  • Manufactured in the United States.