Learn More
MSC Huot End Mill Dispenser, 3 Drawer, 1/8 to 1 Inch, Steel
Catalog No. 5000513260
Encompass
This steel storage unit features three drawers, providing organized storage for industrial-grade end mills. It is designed to accommodate end mills ranging from 1/8 inch to 1 inch in diameter and comes with a tough epoxy powder coating for durability.
  • Accommodates end mills from 1/8 to 1 inch in diameter
  • Equipped with 3 drawers
  • Contains 29 compartments for various sizes
  • Constructed from steel with a durable gray epoxy powder coating
  • Features a stackable design for efficient storage
  • Includes one-piece drawer slides for smooth operation
  • Ideal for managing small inventories of end mills
  • Measures 14.625 in x 7.375 in x 7.75 in (W x D x H)
  • Weighs 10.25 lbs
Catalog No. 50-005-13260 Supplier MSC Supplier No. 01512474
This steel storage unit features three drawers, providing organized storage for industrial-grade end mills. It is designed to accommodate end mills ranging from 1/8 inch to 1 inch in diameter and comes with a tough epoxy powder coating for durability.
  • Accommodates end mills from 1/8 to 1 inch in diameter
  • Equipped with 3 drawers
  • Contains 29 compartments for various sizes
  • Constructed from steel with a durable gray epoxy powder coating
  • Features a stackable design for efficient storage
  • Includes one-piece drawer slides for smooth operation
  • Ideal for managing small inventories of end mills
  • Measures 14.625 in x 7.375 in x 7.75 in (W x D x H)
  • Weighs 10.25 lbs