Learn More
MSC Huot Master Drill Dispenser, Fractional, Letter, Wire Gauge, 159 Compartments, 4-Drawer
Catalog No. 5000533251
Encompass
This master drill dispenser is a 4-drawer steel cabinet designed for organizing fractional, letter, and wire gauge drill bits. It features 159 compartments for efficient storage, ball bearing slides for smooth operation, and a lockable design for security. This dispenser is ideal for laboratories and workshops needing organized access to various drill bit sizes.
  • Holds full packages of jobber or screw machine length drills.
  • Organizes fractional, letter, and wire gauge sizes.
  • Ball bearing slides for easy drawer operation.
  • Lockable with 2 keys.
  • Stackable for larger inventories.
  • 159 compartments.
  • Bottom drawer is empty for additional storage.
  • Steel cabinet with vinyl dividers protects and separates bits.
  • Made in the USA.
Catalog No. 50-005-33251 Supplier MSC Supplier No. 01691153
This master drill dispenser is a 4-drawer steel cabinet designed for organizing fractional, letter, and wire gauge drill bits. It features 159 compartments for efficient storage, ball bearing slides for smooth operation, and a lockable design for security. This dispenser is ideal for laboratories and workshops needing organized access to various drill bit sizes.
  • Holds full packages of jobber or screw machine length drills.
  • Organizes fractional, letter, and wire gauge sizes.
  • Ball bearing slides for easy drawer operation.
  • Lockable with 2 keys.
  • Stackable for larger inventories.
  • 159 compartments.
  • Bottom drawer is empty for additional storage.
  • Steel cabinet with vinyl dividers protects and separates bits.
  • Made in the USA.