Learn More
MSC Huot NC/CNC Collet Storage Rack, Benchtop, 36 Tool Capacity
Catalog No. 5000532840
Encompass
The Huot NC/CNC Collet Storage Rack is designed for efficient organization and storage of collets and toolholders. This benchtop rack has a capacity for 36 tools, including toolholders and various collet sizes, making it suitable for laboratory and workshop environments.
  • Holds 12 toolholders, 12 ER32 collets, and 12 ER16 collets.
  • Benchtop rack for convenient access.
  • Total capacity for 36 tools.
  • Black finish.
  • Well-constructed and space-saving design.
  • Made in the United States.
Catalog No. 50-005-32840 Supplier MSC Supplier No. 53706198
The Huot NC/CNC Collet Storage Rack is designed for efficient organization and storage of collets and toolholders. This benchtop rack has a capacity for 36 tools, including toolholders and various collet sizes, making it suitable for laboratory and workshop environments.
  • Holds 12 toolholders, 12 ER32 collets, and 12 ER16 collets.
  • Benchtop rack for convenient access.
  • Total capacity for 36 tools.
  • Black finish.
  • Well-constructed and space-saving design.
  • Made in the United States.