Learn More
MSC Huot Tap and Drill Dispenser, 3-Drawer, #6-32 to 1/2-20, Steel
Catalog No. 5000529345
Encompass
This 3-drawer tap and drill dispenser is designed to store and organize taps and drills, specifically covering sizes from #6-32 to 1/2-20. Constructed from steel, it provides durable storage for cutting tools.
  • Stores and organizes the correct drill with its corresponding size tap.
  • Holds a minimum of one dozen of each size.
  • Label holders on drawer fronts allow for easy, quick identification and inventory checks.
  • One-piece drawer slide.
  • Stackable with other cutting tool storage cabinets to save space.
  • Finished with a tough epoxy powder coating for durability and protection.
Catalog No. 50-005-29345 Supplier MSC Supplier No. 01512219
This 3-drawer tap and drill dispenser is designed to store and organize taps and drills, specifically covering sizes from #6-32 to 1/2-20. Constructed from steel, it provides durable storage for cutting tools.
  • Stores and organizes the correct drill with its corresponding size tap.
  • Holds a minimum of one dozen of each size.
  • Label holders on drawer fronts allow for easy, quick identification and inventory checks.
  • One-piece drawer slide.
  • Stackable with other cutting tool storage cabinets to save space.
  • Finished with a tough epoxy powder coating for durability and protection.