Learn More
Cayman Chemical (−)-Huperzine A, Purity: ≥98%, CAS: 102518-79-6, 10 mg
SDP
Catalog No. 502968932
Encompass_Preferred
A natural sesquiterpene alkaloid that inhibits AChE in rat cortex in vitro (IC50 = 82 nM) and displays oral AChE inhibitory activity in animals
Catalog No. 50-296-8932 Supplier Cayman Chemical Supplier No. 1462010 MG
A natural sesquiterpene alkaloid that inhibits AChE in rat cortex in vitro (IC50 = 82 nM) and displays oral AChE inhibitory activity in animals