Learn More
Medchemexpress LLC (-)-Huperzine A | 102518-79-6 | 242.32 | 1 ML
SDP
Catalog No. 5000385414
Encompass_Preferred
(-)-Huperzine A is an alkaloid isolated from *Huperzia serrata*, possessing neuroprotective activity. It functions as a potent, highly specific, reversible, and blood-brain barrier penetrant inhibitor of acetylcholinesterase (AChE), with an IC50 of 82 nM. Additionally, it acts as a non-competitive antagonist of the N-methyl-D-aspartate glutamate (NMDA) receptor. This compound is developed for the research of neurodegenerative diseases, including Alzheimer's disease.
  • Alkaloid isolated from *Huperzia serrata*.
  • Exhibits neuroprotective activity.
  • Potent, highly specific, reversible, and blood-brain barrier penetrant inhibitor of acetylcholinesterase (AChE) with an IC50 of 82 nM.
  • Non-competitive antagonist of the N-methyl-D-aspartate glutamate (NMDA) receptor.
  • Developed for research into neurodegenerative diseases, such as Alzheimer's disease.
Catalog No. 50-003-85414 Supplier Medchemexpress LLC Supplier No. HY1738710MM/1ML
(-)-Huperzine A is an alkaloid isolated from *Huperzia serrata*, possessing neuroprotective activity. It functions as a potent, highly specific, reversible, and blood-brain barrier penetrant inhibitor of acetylcholinesterase (AChE), with an IC50 of 82 nM. Additionally, it acts as a non-competitive antagonist of the N-methyl-D-aspartate glutamate (NMDA) receptor. This compound is developed for the research of neurodegenerative diseases, including Alzheimer's disease.
  • Alkaloid isolated from *Huperzia serrata*.
  • Exhibits neuroprotective activity.
  • Potent, highly specific, reversible, and blood-brain barrier penetrant inhibitor of acetylcholinesterase (AChE) with an IC50 of 82 nM.
  • Non-competitive antagonist of the N-methyl-D-aspartate glutamate (NMDA) receptor.
  • Developed for research into neurodegenerative diseases, such as Alzheimer's disease.