Learn More
Medchemexpress LLC (-)-Huperzine A | 102518-79-6 | 242.32 | 100 MG
SDP
Catalog No. 5000385013
Encompass_Preferred
(-)-Huperzine A is an alkaloid isolated from Huperzia serrata, possessing neuroprotective activity. It is a potent, highly specific, reversible, and blood-brain barrier penetrant inhibitor of acetylcholinesterase (AChE), with an IC50 of 82 nM. This compound also acts as a non-competitive antagonist of the N-methyl-D-aspartate glutamate (NMDA) receptor and is developed for the research of neurodegenerative diseases, including Alzheimer's disease.
  • Alkaloid isolated from Huperzia serrata
  • Possesses neuroprotective activity
  • Potent, highly specific, reversible, and blood-brain barrier penetrant inhibitor of acetylcholinesterase (AChE) (IC50 = 82 nM)
  • Non-competitive antagonist of N-methyl-D-aspartate glutamate (NMDA) receptor
  • Developed for research into neurodegenerative diseases, such as Alzheimer's disease
Catalog No. 50-003-85013 Supplier Medchemexpress LLC Supplier No. HY17387100MG
(-)-Huperzine A is an alkaloid isolated from Huperzia serrata, possessing neuroprotective activity. It is a potent, highly specific, reversible, and blood-brain barrier penetrant inhibitor of acetylcholinesterase (AChE), with an IC50 of 82 nM. This compound also acts as a non-competitive antagonist of the N-methyl-D-aspartate glutamate (NMDA) receptor and is developed for the research of neurodegenerative diseases, including Alzheimer's disease.
  • Alkaloid isolated from Huperzia serrata
  • Possesses neuroprotective activity
  • Potent, highly specific, reversible, and blood-brain barrier penetrant inhibitor of acetylcholinesterase (AChE) (IC50 = 82 nM)
  • Non-competitive antagonist of N-methyl-D-aspartate glutamate (NMDA) receptor
  • Developed for research into neurodegenerative diseases, such as Alzheimer's disease