Learn More
Selleck Chemical LLC (-)-Huperzine A (HupA) S2251-2mg
SDP
Catalog No. 503112725
Encompass_Preferred
(-)-Huperzine A is a potent highly specific and reversible inhibitor of acetylcholinesterase (AChE) with Ki of 7 nM exhibiting 200-fold more selectivity for G4 AChE over G1 AChE Also acts as an NMDA receptor antagonist Phase 4
Catalog No. 50-311-2725 Supplier Selleck Chemical LLC Supplier No. S22512MG
(-)-Huperzine A is a potent highly specific and reversible inhibitor of acetylcholinesterase (AChE) with Ki of 7 nM exhibiting 200-fold more selectivity for G4 AChE over G1 AChE Also acts as an NMDA receptor antagonist Phase 4