Learn More
Enzo Life Sciences Huperzine A (5mg). CAS: 120786-18-7
SDP
Supplier:  Enzo Life Sciences ALX550064M005
Encompass_Preferred
Racemic modification of (-)-Huperzine A (ALX-550-065) with acetylcholinesterase inhibitor activity. Has a weaker biological activity than (-)Huperzine A. Neurotoxic. Alternative name: (+/-)-Selagine. Purity: ≥95%. Solubility: Soluble in dilute aqueous acids or chloroform. Long Term Storage: +4°C.
Catalog No. 50-200-9472