Learn More
Selleck Chemical LLC (-)-Huperzine A (HupA) 10mg 102518-79-6
SDP
Supplier:  Selleck Chemical LLC S225110MG
Encompass_Preferred
(-)-Huperzine A is a potent, highly specific and reversible inhibitor of acetylcholinesterase (AChE) with Ki of 7 nM, exhibiting 200-fold more selectivity for G4 AChE over G1 AChE. Also acts as an NMDA receptor antagonist. Phase 4. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-706-4