Learn More
Medchemexpress LLC Huperzine B | 103548-82-9 | 256.34 | 20 MG
SDP
Catalog No. 5000415886
Encompass_Preferred
Huperzine B is a Lycopodium alkaloid isolated from *Huperzia serrata* and acts as a highly selective acetylcholinesterase (AChE) inhibitor. It can be used to improve Alzheimer's disease.
  • Lycopodium alkaloid isolated from *Huperzia serrata*.
  • Highly selective acetylcholinesterase (AChE) inhibitor.
  • Can be used to improve Alzheimer's disease.
  • Purity: 98.73%.
Catalog No. 50-004-15886 Supplier Medchemexpress LLC Supplier No. HYN204320MG
Huperzine B is a Lycopodium alkaloid isolated from *Huperzia serrata* and acts as a highly selective acetylcholinesterase (AChE) inhibitor. It can be used to improve Alzheimer's disease.
  • Lycopodium alkaloid isolated from *Huperzia serrata*.
  • Highly selective acetylcholinesterase (AChE) inhibitor.
  • Can be used to improve Alzheimer's disease.
  • Purity: 98.73%.