Learn More
Medchemexpress LLC Huperzine B | 103548-82-9 | MFCD01716207 | 98.7% | 256.34 g/mol | C16H20N2O | 10 MG
SDP
Catalog No. 5000295662
Encompass_Preferred
Huperzine B is a Lycopodium alkaloid and a selective acetylcholinesterase (AChE) inhibitor used as a research reagent. It is supplied as a purified white to off-white solid with high purity and is suitable for biochemical and pharmacological studies.
  • Selective acetylcholinesterase inhibitor.
  • High purity (98.7%).
  • White to off-white solid form.
  • Available in milligram quantities and in DMSO solution for assays.
  • Suitable for biochemical and pharmacological research applications.
Catalog No. 50-002-95662 Supplier Medchemexpress LLC Supplier No. HYN204310MG
Huperzine B is a Lycopodium alkaloid and a selective acetylcholinesterase (AChE) inhibitor used as a research reagent. It is supplied as a purified white to off-white solid with high purity and is suitable for biochemical and pharmacological studies.
  • Selective acetylcholinesterase inhibitor.
  • High purity (98.7%).
  • White to off-white solid form.
  • Available in milligram quantities and in DMSO solution for assays.
  • Suitable for biochemical and pharmacological research applications.