Learn More
Proteintech Group Inc HuR Polyclonal antibody
SDP
Supplier:  Proteintech Group Inc 119101AP150UL
Encompass_Preferred
The HuR antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human HuR. This antibody recognizes human, mouse, rat antigen. The HuR antibody has been validated for the following applications: FC, IF, IHC, IP, WB, ELISA analysis.
Catalog No. 50-225-6378