Learn More
Eurofins Discoverx huRsk1/MAPKAP Kinase 1a, active, 10 µg. (14-509)
Supplier:  Eurofins Discoverx 14509
Encompass
N-terminal 6xHis-tagged recombinant, human full length RSK1. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8891