Learn More
Refrigiwear Inc. HUSKY FREEZER GLOVES
SDP
Catalog No. NC3868088
Encompass_Preferred
Made for hard work in the cold, the Husky Freezer Gloves offers warmth and dexterity down to -15°F (-26°C).
Catalog No. NC3868088 Supplier Refrigiwear Inc. Supplier No. 2115RBCHLAR
Made for hard work in the cold, the Husky Freezer Gloves offers warmth and dexterity down to -15°F (-26°C).