Learn More
Promocell GmbH PromoCell HUVEC-c 500,000 Human Umbilical Vein Endothelial Cells, single donor, cryopreserved
Catalog No. 50305962
Encompass
Primary Human Umbilical Vein Endothelial Cells (HUVEC) are isolated from the vein of the umbilical cord and are commonly used for physiological and pharmacological investigations, such as macromolecule transport, blood coagulation, angiogenesis, and fibrinolysis.PromoCell offers HUVEC isolated in standard Endothelial Cell Growth Medium. The cells are supplied either from single donors or from pooled donors (from up to four different umbilical cords).In addition, HUVEC are available which are pre-screened for VEGF (vascular endothelial growth factor) response. VEGF is an important signaling protein involved in both vasculogenesis and angiogenesis. In vitro, VEGF has been shown to stimulate endothelial cell mitogenesis, cell migration, sprouting, and microvascular permeability.
Catalog No. 50-305-962 Supplier Promocell GmbH Supplier No. C12200
Primary Human Umbilical Vein Endothelial Cells (HUVEC) are isolated from the vein of the umbilical cord and are commonly used for physiological and pharmacological investigations, such as macromolecule transport, blood coagulation, angiogenesis, and fibrinolysis.PromoCell offers HUVEC isolated in standard Endothelial Cell Growth Medium. The cells are supplied either from single donors or from pooled donors (from up to four different umbilical cords).In addition, HUVEC are available which are pre-screened for VEGF (vascular endothelial growth factor) response. VEGF is an important signaling protein involved in both vasculogenesis and angiogenesis. In vitro, VEGF has been shown to stimulate endothelial cell mitogenesis, cell migration, sprouting, and microvascular permeability.