Learn More
Promocell GmbH PromoCell HUVEC-c 500,000 Human Umbilical Vein Endothelial Cells, isolated in Growth Medium 2, pooled, cryopreserved
Supplier:  Promocell GmbH C12208
Encompass
Primary Human Umbilical Vein Endothelial Cells (HUVEC) are isolated from the vein of the umbilical cord and are commonly used for physiological and pharmacological investigations, such as macromolecule transport, blood coagulation, angiogenesis, and fibrinolysis.PromoCell offers HUVEC isolated in Endothelial Cell Growth Medium 2 which is free of ECGS and contains additional IGF and VEGF. The cells are supplied either from single donors or from pooled donors (from up to four different umbilical cords).
Catalog No. NC1104944