Learn More
Lonza Walkersville HUVEC – Human Umbilical Vein Endothelial Cells, Single Donor, in EGM-2
Supplier:  Lonza Walkersville C2517A
Encompass
Lonza's Cryopreserved HUVECs are shipped as frozen primaries. They are guaranteed to express CD31/105 and survive in culture through 15* population doublings (*except HUVEC-XL guaranteed through 5 PD).All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis is provided for each cell lot purchased.Benefits:Guaranteed through 15 population doublings Test negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsA Certificate of Analysis is provided for each cell lot purchased Applications:Wound healingAngiogenesisTissue engineeringInflammationOncologyPharmacologyVascular modelingTransfection Content:1 x Cryopreserved ampule of HUVEC containing ≥ 500,000 cells
Catalog No. NC9911820