Learn More
Lonza Walkersville HUVEC – Human Umbilical Vein Endothelial Cells, Pooled, in EGM
Supplier:  Lonza Walkersville CC2519
Encompass
Lonza's Cryopreserved HUVECs are shipped as frozen primaries. They are guaranteed to express CD31/105 and survive in culture through 15* population doublings (*except HUVEC-XL guaranteed through 5 PD).All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis is provided for each cell lot purchased.Benefits:Guaranteed through 15 population doublings Single or pooled donors available All cells test negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsCertificate of Analysis provided for every cell lot Applications:Would healingAngiogenesisTissue engineeringInflammationOncologyPharmacologyVascular modelingTransfection Content:1 x Cryopreserved ampule HUVEC (pooled donor) containing ≥500,000 cells
Catalog No. NC1835181