Learn More
Lonza Walkersville HUVEC – Human Umbilical Vein Endothelial Cells, Single Donor, in EGM
Catalog No. NC9254075
Encompass
Lonza's Cryopreserved HUVECs are shipped as frozen primaries. They are guaranteed to express CD31/105 and survive in culture through 15* population doublings (*except HUVEC-XL guaranteed through 5 PD).All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis is provided for each cell lot purchased.Benefits:Guaranteed through 15 population doublings Pooled or single donors available All cells test negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsCertificate of Analysis provided for every cell lot Applications:Would healingAngiogenesisTissue engineeringInflammationOncologyPharmacologyVascular modelingTransfection
This item is not returnable. View return policy
Catalog No. NC9254075 Supplier Lonza Walkersville Supplier No. CC2517
Add to Cart
Add to Cart
Lonza's Cryopreserved HUVECs are shipped as frozen primaries. They are guaranteed to express CD31/105 and survive in culture through 15* population doublings (*except HUVEC-XL guaranteed through 5 PD).All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis is provided for each cell lot purchased.Benefits:Guaranteed through 15 population doublings Pooled or single donors available All cells test negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsCertificate of Analysis provided for every cell lot Applications:Would healingAngiogenesisTissue engineeringInflammationOncologyPharmacologyVascular modelingTransfection