Learn More
Lonza Walkersville HUVEC – Human Umbilical Vein Endothelial Cells, Pooled, in EGM-2, Prescreened for Angiogenesis
Catalog No. NC0565869
Encompass
Lonza's Cryopreserved HUVECs are shipped as frozen primaries. They are guaranteed through 15* population doublings (*except HUVEC-XL guaranteed through 5 PD), express CD31/105, von Williebrand Factor VIII, and are positive for acetylated low density lipoprotein uptake. All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis is provided for each cell lot purchased.Benefits:Guaranteed through 15 population doublings All cells test negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsCertificate of Analysis provided for every cell lot Applications:Would healingAngiogenesisTissue engineeringInflammationOncologyPharmacologyVascular modelingTransfection Content:1 x Cryopreserved ampule HUVEC containing ≥ 500,000 cells/vial
Catalog No. NC0565869 Supplier Lonza Walkersville Supplier No. C2519AS
Lonza's Cryopreserved HUVECs are shipped as frozen primaries. They are guaranteed through 15* population doublings (*except HUVEC-XL guaranteed through 5 PD), express CD31/105, von Williebrand Factor VIII, and are positive for acetylated low density lipoprotein uptake. All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis is provided for each cell lot purchased.Benefits:Guaranteed through 15 population doublings All cells test negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsCertificate of Analysis provided for every cell lot Applications:Would healingAngiogenesisTissue engineeringInflammationOncologyPharmacologyVascular modelingTransfection Content:1 x Cryopreserved ampule HUVEC containing ≥ 500,000 cells/vial