Learn More
Lonza Walkersville HUVEC – Human Umbilical Vein Endothelial Cells, Pooled, in EGM-2
Catalog No. NC9946677
Encompass
Lonza's Cryopreserved HUVECs are shipped as frozen primaries. They are guaranteed to express CD31/105 and survive in culture through 15* population doublings (*except HUVEC-XL guaranteed through 5 PD).All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis is provided for each cell lot purchased.Benefits:Guaranteed through 15 population doublings Test negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsA Certificate of Analysis is provided for each cell lot purchased Applications:Wound healingAngiogenesisTissue engineeringInflammationOncologyPharmacologyVascular modelingTransfectionContent:1 x Cryopreserved ampule of HUVEC containing ≥ 500,000 cells
This item is not returnable. View return policy
Catalog No. NC9946677 Supplier Lonza Walkersville Supplier No. C2519A
Lonza's Cryopreserved HUVECs are shipped as frozen primaries. They are guaranteed to express CD31/105 and survive in culture through 15* population doublings (*except HUVEC-XL guaranteed through 5 PD).All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis is provided for each cell lot purchased.Benefits:Guaranteed through 15 population doublings Test negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsA Certificate of Analysis is provided for each cell lot purchased Applications:Wound healingAngiogenesisTissue engineeringInflammationOncologyPharmacologyVascular modelingTransfectionContent:1 x Cryopreserved ampule of HUVEC containing ≥ 500,000 cells