Learn More
Carbosynth LLC. Huwentoxin-IV (0.1mg vial), 1ea, Sequence originally discovered from the venom of the Chinese bird spider, Selenocosmia huwena., Disulfide bonds: Disulfide bonds between Cys2- Cys17, Cys9- Cys24, and Cys16- Cys31, MW: 4106.8, Molecular formula: C174H278N52O51S6
SDP
Supplier:  Carbosynth LLC. PCB4455S0.1MG
Encompass_Preferred
Synonyms: HWTX-IV, Glu-Cys-Leu-Glu-Ile-Phe-Lys-Ala-Cys-Asn-Pro-Ser-Asn-Asp-Gln-Cys-Cys-Lys-Ser-Ser-Lys-Leu-Val-Cys-Ser-Arg-Lys-Thr-Arg-Trp-Cys-Lys-Tyr-Gln-Ile-NH2, Application: Neuronal Tetrodotoxin-Sensitive Na+- Channel Blocker, Salt Form:, Trifluoroacetate Salt, Storage Temperature: -20°C, References: K. Peng, Q. Shu, Z. Liu, and S. Liang, J. Biol. Chem., 277, 47564 (2002). (Original), J. Diao, Y. Lin, J. Tang, and S. Liang, Toxicon, 42, 715 (2003). (cDNA Seq), Y. Xiao, J.-P. Bingham, W. Zhu, E. Moczydlowski, S. Liang, and T.R. Cummins, J. Biol. Chem., 283, 27300 (2008). (Pharmacol.)
Catalog No. 50-168-6263