Learn More
Medchemexpress LLC Huwentoxin-IV TFA | 105856-78-4 | 99.04% | 4099.71 | 5 MG
SDP
Catalog No. 5000423613
Encompass_Preferred
Huwentoxin-IV TFA is a highly potent and selective blocker of neuronal voltage-gated sodium channels. It specifically inhibits NaV1.7 activity in human DRG neurons.
  • Potent and selective sodium channel blocker
  • Inhibits neuronal NaV1.7, NaV1.2, NaV1.3, and NaV1.4 channels
  • IC50 of 2.9 nM for inhibiting NaV1.7-mediated currents
  • Reduces NaV1.7 current amplitude by 50% at 0.5 nM
Catalog No. 50-004-23613 Supplier Medchemexpress LLC Supplier No. HYP1220A5MG
Huwentoxin-IV TFA is a highly potent and selective blocker of neuronal voltage-gated sodium channels. It specifically inhibits NaV1.7 activity in human DRG neurons.
  • Potent and selective sodium channel blocker
  • Inhibits neuronal NaV1.7, NaV1.2, NaV1.3, and NaV1.4 channels
  • IC50 of 2.9 nM for inhibiting NaV1.7-mediated currents
  • Reduces NaV1.7 current amplitude by 50% at 0.5 nM