Learn More
Medchemexpress LLC Huwentoxin-IV TFA | 526224-73-7 | 4106.78 | 500 UG
SDP
Catalog No. 5000423612
Encompass_Preferred
Huwentoxin-IV is a potent and selective sodium channel blocker. It inhibits neuronal Nav1.7, Nav1.2, Nav1.3, and Nav1.4, with preferential blocking of peripheral nerve subtype Nav1.7. This compound has also demonstrated analgesic effects in animal models of inflammatory and neuropathic pain.
  • Potent and selective sodium channel blocker
  • Inhibits neuronal Nav1.7, Nav1.2, Nav1.3, and Nav1.4
  • Preferentially blocks peripheral nerve subtype Nav1.7
  • Binds to neurotoxin receptor site 4
  • Analgesic effects in animal models of inflammatory and neuropathic pain
Catalog No. 50-004-23612 Supplier Medchemexpress LLC Supplier No. HYP1220A500UG
Huwentoxin-IV is a potent and selective sodium channel blocker. It inhibits neuronal Nav1.7, Nav1.2, Nav1.3, and Nav1.4, with preferential blocking of peripheral nerve subtype Nav1.7. This compound has also demonstrated analgesic effects in animal models of inflammatory and neuropathic pain.
  • Potent and selective sodium channel blocker
  • Inhibits neuronal Nav1.7, Nav1.2, Nav1.3, and Nav1.4
  • Preferentially blocks peripheral nerve subtype Nav1.7
  • Binds to neurotoxin receptor site 4
  • Analgesic effects in animal models of inflammatory and neuropathic pain