Learn More
Electron Microscopy Sciences Huygenian Eyepiece, X7
SDP
Catalog No. 5025831
Encompass_Preferred
Focusing eyepiece composed of two plano convex lenses, separated by an air space. The image plane is between the two lenses. The top lens is adjusted to bring the graticule into focus.
Catalog No. 50-258-31 Supplier Electron Microscopy Sciences Supplier No. 67990F7
Focusing eyepiece composed of two plano convex lenses, separated by an air space. The image plane is between the two lenses. The top lens is adjusted to bring the graticule into focus.