Learn More
Cayman Chemical HValProProOHtrIfluoroac 10mg
SDP
Catalog No. 502939772
Encompass_Preferred
A peptide ACE inhibitor (IC50 = 9 µM); inhibits acetylcholine-induced NO production in HUVECs at 1 µM; induces vasorelaxation in precontracted isolated rat aortic rings; decreases systolic blood pressure in spontaneously hypertensive rats at 0.3 mg/kg; dietary administration reduces plasma levels of total cholesterol, HDL-cholesterol, and triglycerides, expression of Il6 and Il1b, and aortic arch intimal thickening and atherosclerotic plaque formation in ApoE-/- mice
Catalog No. 50-293-9772 Supplier Cayman Chemical Supplier No. 2889810 MG
A peptide ACE inhibitor (IC50 = 9 µM); inhibits acetylcholine-induced NO production in HUVECs at 1 µM; induces vasorelaxation in precontracted isolated rat aortic rings; decreases systolic blood pressure in spontaneously hypertensive rats at 0.3 mg/kg; dietary administration reduces plasma levels of total cholesterol, HDL-cholesterol, and triglycerides, expression of Il6 and Il1b, and aortic arch intimal thickening and atherosclerotic plaque formation in ApoE-/- mice