Learn More
Cayman Chemical H-Val-Pro-Pro-OH (trifluoroactetate salt), Purity: ≥95%, 50 mg
SDP
Catalog No. 502940546
Encompass_Preferred
A peptide ACE inhibitor (IC50 = 9 µM); inhibits acetylcholine-induced NO production in HUVECs at 1 µM; induces vasorelaxation in precontracted isolated rat aortic rings; decreases systolic blood pressure in spontaneously hypertensive rats at 0.3 mg/kg; dietary administration reduces plasma levels of total cholesterol, HDL-cholesterol, and triglycerides, expression of Il6 and Il1b, and aortic arch intimal thickening and atherosclerotic plaque formation in ApoE-/- mice
Catalog No. 50-294-0546 Supplier Cayman Chemical Supplier No. 2889850 MG
A peptide ACE inhibitor (IC50 = 9 µM); inhibits acetylcholine-induced NO production in HUVECs at 1 µM; induces vasorelaxation in precontracted isolated rat aortic rings; decreases systolic blood pressure in spontaneously hypertensive rats at 0.3 mg/kg; dietary administration reduces plasma levels of total cholesterol, HDL-cholesterol, and triglycerides, expression of Il6 and Il1b, and aortic arch intimal thickening and atherosclerotic plaque formation in ApoE-/- mice