Learn More
Medchemexpress LLC hVEGF-IN-1 | 1637443-98-1 | 99.3% | 581.75 g/mol | C34H43N7O2 | 100 MG
SDP
Catalog No. 5000248325
Encompass_Preferred
hVEGF-IN-1 is a quinazoline-derived small-molecule research reagent that inhibits VEGF-A expression by binding G-rich sequences within the IRES-A element and destabilizing G-quadruplex structures. It has been reported to reduce VEGF-A protein levels, hinder tumor cell migration, and suppress tumor growth in cellular assays.
  • Binds G-rich IRES-A sequence and destabilizes G-quadruplex structures.
  • Reduces VEGF-A protein expression in cellular assays.
  • Inhibits tumor cell migration and growth in vitro.
  • High chemical purity suitable for research applications.
  • Available in multiple solid and solution package sizes for experimental flexibility.
Catalog No. 50-002-48325 Supplier Medchemexpress LLC Supplier No. HY101931100MG
hVEGF-IN-1 is a quinazoline-derived small-molecule research reagent that inhibits VEGF-A expression by binding G-rich sequences within the IRES-A element and destabilizing G-quadruplex structures. It has been reported to reduce VEGF-A protein levels, hinder tumor cell migration, and suppress tumor growth in cellular assays.
  • Binds G-rich IRES-A sequence and destabilizes G-quadruplex structures.
  • Reduces VEGF-A protein expression in cellular assays.
  • Inhibits tumor cell migration and growth in vitro.
  • High chemical purity suitable for research applications.
  • Available in multiple solid and solution package sizes for experimental flexibility.