Learn More
ACROBiosystems ACROBiosystems Human HVEM / TNFRSF14 Protein, His Tag
Supplier:  ACROBiosystems HVMH52E91MG
Encompass_Preferred
Recombinant Protein;1MG;Human HVEM, His Tag (HVM-H52E9) is expressed from human 293 cells (HEK293). It contains AA Leu 39 - Val 202 (Accession # Q92956-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.2 kDa. The protein migrates as 33-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LIGHT, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. LIT-H5256) at 2 μg/mL (100 μL/well) can bind Human HVEM, His Tag (Cat. No. HVM-H52E9) with a linear range of 0.04-0.6 μg/mL (QC tested).;Immobilized Human HVEM, His Tag (Cat. No. HVM-H52E9) at 5 μg/mL (100 μL/well) can bind Human BTLA, Fc Tag (Cat. No. BTA-H5256) with a linear range of 0.2-2.5 μg/mL (Routinely tested).
Catalog No. 50-195-7292