Learn More
Sino Biological Recombinant Human HVEM Protein (His Tag), Amine-Labeled Biotinylated, HPLC-verified
Supplier:  Sino Biological 10334H08HB100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Val202) of human HVEM (NP_003811.2) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6176