Learn More
MSC Hoover Commercial Battery Charger, Fast, 40V
Catalog No. 502836701
Encompass
This Hoover Commercial HVRPWR 40V Fast Charger efficiently charges and maintains 40V commercial batteries. It integrates advanced safety features like temperature monitoring, sleep mode, surge protection, and auto charging/discharging termination for optimal battery performance and longevity.
  • Monitors and regulates battery temperature to prevent overheating.
  • Activates charging at optimal temperature.
  • Protects battery cells with surge protection.
  • Regulates maximum charge time and terminates charging/discharging automatically.
  • Includes LED status indicators.
  • UL certified for safety.
  • Allows easy battery removal.
  • Designed for commercial applications.
Catalog No. 50-283-6701 Supplier MSC Supplier No. 76449354
This Hoover Commercial HVRPWR 40V Fast Charger efficiently charges and maintains 40V commercial batteries. It integrates advanced safety features like temperature monitoring, sleep mode, surge protection, and auto charging/discharging termination for optimal battery performance and longevity.
  • Monitors and regulates battery temperature to prevent overheating.
  • Activates charging at optimal temperature.
  • Protects battery cells with surge protection.
  • Regulates maximum charge time and terminates charging/discharging automatically.
  • Includes LED status indicators.
  • UL certified for safety.
  • Allows easy battery removal.
  • Designed for commercial applications.