Learn More
MSC TPI Industrial Circulator Fan, Steel, 30 in., 1/2 HP, 1 ea.
Catalog No. 5000507181
Encompass
The Heavy Duty 1/2 HP Stationary Circulator Head is a robust industrial fan designed for reliable air circulation. This fully-assembled unit features a durable steel construction with a silver/gray powder-coated finish and a black painted motor housing. Engineered for high performance, it utilizes a 1/2 HP direct-drive, totally enclosed air-over (TEAO) motor to deliver consistent airflow.
  • Features a 30 in. diameter aluminum 3-paddle blade system with a steel hub and cast aluminum spider connections.
  • Equipped with a pull-chain switch and a 12 ft. SJT-type cord with a grounded plug for plug-and-play operation.
  • Includes self-locking retaining clips to secure front and rear grills, meeting OSHA safety requirements.
  • Operates at two speeds with high/low settings.
  • Designed for non-oscillating, stationary mounting.
  • Backed by a 5-year warranty.
Catalog No. 50-005-07181 Supplier MSC Supplier No. 77994424
The Heavy Duty 1/2 HP Stationary Circulator Head is a robust industrial fan designed for reliable air circulation. This fully-assembled unit features a durable steel construction with a silver/gray powder-coated finish and a black painted motor housing. Engineered for high performance, it utilizes a 1/2 HP direct-drive, totally enclosed air-over (TEAO) motor to deliver consistent airflow.
  • Features a 30 in. diameter aluminum 3-paddle blade system with a steel hub and cast aluminum spider connections.
  • Equipped with a pull-chain switch and a 12 ft. SJT-type cord with a grounded plug for plug-and-play operation.
  • Includes self-locking retaining clips to secure front and rear grills, meeting OSHA safety requirements.
  • Operates at two speeds with high/low settings.
  • Designed for non-oscillating, stationary mounting.
  • Backed by a 5-year warranty.