Learn More
MSC Rochester Midland Corporation Powdered Cleaner and Degreaser, All-Metal Safe, Non-Caustic, 400 lb
Catalog No. 502835777
Encompass
PA 290 Powdered Cleaner and Degreaser is an all-metal-safe cleaner designed to effectively remove a broad spectrum of soils, including baked and charred oils, grease, shop soils, and carbon. This heavy-duty degreaser is non-caustic, making it safe for all metals and suitable for a wide range of cleaning functions. It offers superior grease penetration and soil suspension, ensuring a free-rinsing and non-filming finish. Its concentrated formula provides a low use cost, and it dissolves totally, easily eliminating waste.
  • Non-caustic and safe for all metals
  • Superior grease penetration and soil suspension
  • Free-rinsing and non-filming formula
  • Concentrated for low use cost
  • Versatile, effective over a wide range of cleaning functions
  • Dissolves totally, easily eliminating waste
Catalog No. 50-283-5777 Supplier MSC Supplier No. 54708920
PA 290 Powdered Cleaner and Degreaser is an all-metal-safe cleaner designed to effectively remove a broad spectrum of soils, including baked and charred oils, grease, shop soils, and carbon. This heavy-duty degreaser is non-caustic, making it safe for all metals and suitable for a wide range of cleaning functions. It offers superior grease penetration and soil suspension, ensuring a free-rinsing and non-filming finish. Its concentrated formula provides a low use cost, and it dissolves totally, easily eliminating waste.
  • Non-caustic and safe for all metals
  • Superior grease penetration and soil suspension
  • Free-rinsing and non-filming formula
  • Concentrated for low use cost
  • Versatile, effective over a wide range of cleaning functions
  • Dissolves totally, easily eliminating waste