Learn More
MSC Rochester Midland Corporation Cleaner & Degreaser, 1 gal, Concentrated, Lemon
Catalog No. 502839785
Encompass
Rochester Midland Corporation's Cleaner & Degreaser is a highly concentrated liquid solution designed for effective removal of grease and encrusted soil. It is versatile and can be used in various applications like soak tanks, spray, and mop. This non-explosive, non-flammable formula is also low in toxicity.
  • Non-explosive, non-flammable, low toxicity
  • Versatile for various cleaning needs
  • Contains powerful synthetic detergents, wetting agent, glycol ether solvent, and alkaline builders
  • Economical to use
  • Highly concentrated for effective cleaning
Catalog No. 50-283-9785 Supplier MSC Supplier No. 92078021
Rochester Midland Corporation's Cleaner & Degreaser is a highly concentrated liquid solution designed for effective removal of grease and encrusted soil. It is versatile and can be used in various applications like soak tanks, spray, and mop. This non-explosive, non-flammable formula is also low in toxicity.
  • Non-explosive, non-flammable, low toxicity
  • Versatile for various cleaning needs
  • Contains powerful synthetic detergents, wetting agent, glycol ether solvent, and alkaline builders
  • Economical to use
  • Highly concentrated for effective cleaning