Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Zep Powerplex Laundry Detergent, Industrial, Liquid, Blue, 20 gal

Catalog No. 502819971
Encompass

Zep Powerplex is an industrial-grade liquid laundry detergent designed for effective cleaning. This blue liquid detergent has a characteristic odor and is supplied in a 20-gallon drum.

  • Liquid form for easy dispensing
  • Characteristic scent
  • Container capacity of 20 gal
  • Has a pH range of 8 to 8.5
  • Density of 1.020 g/cm³
  • Soluble in water
  • Kinematic viscosity of 110.9 mm²/s at 20 °C
  • Combustible liquid with a flash point of 68 °C

Catalog No. 50-281-9971 Supplier MSC Supplier No. 17596412
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zep Powerplex is an industrial-grade liquid laundry detergent designed for effective cleaning. This blue liquid detergent has a characteristic odor and is supplied in a 20-gallon drum.

  • Liquid form for easy dispensing
  • Characteristic scent
  • Container capacity of 20 gal
  • Has a pH range of 8 to 8.5
  • Density of 1.020 g/cm³
  • Soluble in water
  • Kinematic viscosity of 110.9 mm²/s at 20 °C
  • Combustible liquid with a flash point of 68 °C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.