Learn More
Cole-Parmer Cole-Parmer Heavy-Duty Polypropylene Carboy with Shoulder Handle, 5 L
Catalog No. NC0217328 Shop All Cole-Parmer Products
Encompass_Preferred
Graduated in descending 1 L increments to deliver liquids easily. Octagonal shape increases rigidity when full. Offset neck makes filling and pouring a snap. Molded shoulder handle sealed at ends to prevent contamination. Maximum autoclavable temperature is 275F.
Catalog No. NC0217328 Supplier Cole-Parmer Supplier No. 6250700
Graduated in descending 1 L increments to deliver liquids easily. Octagonal shape increases rigidity when full. Offset neck makes filling and pouring a snap. Molded shoulder handle sealed at ends to prevent contamination. Maximum autoclavable temperature is 275F.