Learn More
MSC Sprayon Degreaser, Heavy-Duty, Citrus, Liquid, CD757 Series, 55 gal
Catalog No. 502868187
Encompass
The Sprayon CD757 Citrus Degreaser is a biodegradable, heavy-duty, multi-purpose liquid formula designed for effective removal of grime, dirt, grease, and oil. It is suitable for diverse applications in general maintenance, public facilities, and as a surface preparation cleaner before painting.
  • Biodegradable formula
  • Heavy-duty, multi-purpose cleaner and graffiti remover
  • Removes grimy dirt, grease, and oil
  • Pleasant citrus scent
  • Suitable for various surfaces including wood, ceramic tile, and some plastics
  • Dielectric strength of 40,400 V
  • NSF registered C1 rated for general non-processing areas
  • No chlorinated solvents, CFCs, or HCFCs
  • Ideal as a cleaner prior to painting
Catalog No. 50-286-8187 Supplier MSC Supplier No. 30072540
Add to Cart
Add to Cart
Request a Quote
The Sprayon CD757 Citrus Degreaser is a biodegradable, heavy-duty, multi-purpose liquid formula designed for effective removal of grime, dirt, grease, and oil. It is suitable for diverse applications in general maintenance, public facilities, and as a surface preparation cleaner before painting.
  • Biodegradable formula
  • Heavy-duty, multi-purpose cleaner and graffiti remover
  • Removes grimy dirt, grease, and oil
  • Pleasant citrus scent
  • Suitable for various surfaces including wood, ceramic tile, and some plastics
  • Dielectric strength of 40,400 V
  • NSF registered C1 rated for general non-processing areas
  • No chlorinated solvents, CFCs, or HCFCs
  • Ideal as a cleaner prior to painting