Learn More
MSC CRC Heavy Duty Degreaser, 55 Gal, Liquid, Citrus Scent
Catalog No. 502836527
Encompass
This liquid, industrial strength cleaner and degreaser with a citrus scent is formulated with d-Limonene solvent. It is suitable for a wide range of applications, including air tools, automatic drip machines, bearings, chains, clutches, compressors, floor scrubbers, generators, mechanical brakes, molds, motors, power washers, presses, rollers, and wire ropes.
  • Liquid form
  • 55 gal drum container
  • Citrus scent
  • Removes oil and ink
  • Not a disinfectant
  • Primary chemical is soybean oil
  • NSF K1 registered and RoHS compliant
  • Industrial strength cleaner and degreaser formula
  • Low VOC content: 3.25 g/L (at dilution) and 0.4%
Catalog No. 50-283-6527 Supplier MSC Supplier No. 58181017
This liquid, industrial strength cleaner and degreaser with a citrus scent is formulated with d-Limonene solvent. It is suitable for a wide range of applications, including air tools, automatic drip machines, bearings, chains, clutches, compressors, floor scrubbers, generators, mechanical brakes, molds, motors, power washers, presses, rollers, and wire ropes.
  • Liquid form
  • 55 gal drum container
  • Citrus scent
  • Removes oil and ink
  • Not a disinfectant
  • Primary chemical is soybean oil
  • NSF K1 registered and RoHS compliant
  • Industrial strength cleaner and degreaser formula
  • Low VOC content: 3.25 g/L (at dilution) and 0.4%