Learn More
MSC O-Cedar Floor Sweep, 18 in Wide, Polypropylene Bristle, 6/Pk.
Catalog No. 502816019
Encompass
These heavy-duty floor sweeps are designed for scrubbing both wet and dry surfaces. Each sweep features a 3 1/4 in crimped red polypropylene fill brush that resists acids, oils, fungi, and bacteria, making them suitable for food service and industrial settings. The durable foam block is hygienic, non-absorbent, and includes two threaded holes for handles.
  • Includes 6 sweeps
  • Crimped red polypropylene fill brush is ideal for wet or dry scrubbing
  • Solvent and grease resistant
  • Suitable for food service and industrial use
  • Handles and braces sold separately
  • Made in the USA
Catalog No. 50-281-6019 Supplier MSC Supplier No. 30322499
These heavy-duty floor sweeps are designed for scrubbing both wet and dry surfaces. Each sweep features a 3 1/4 in crimped red polypropylene fill brush that resists acids, oils, fungi, and bacteria, making them suitable for food service and industrial settings. The durable foam block is hygienic, non-absorbent, and includes two threaded holes for handles.
  • Includes 6 sweeps
  • Crimped red polypropylene fill brush is ideal for wet or dry scrubbing
  • Solvent and grease resistant
  • Suitable for food service and industrial use
  • Handles and braces sold separately
  • Made in the USA