Learn More
Electron Microscopy Sciences EMS Heavy Pattern Dissecting Knife with 40 mm rule, 13" OAL
SDP
Catalog No. 501931584
Encompass_Preferred
This extra thick, rigid stainless steel blade is perfect for cutting hard tissue. Features a thick tapered edge with an optional 40 mm imprinted metric ruler. The top edge is 0.062" (1.6 mm) thick and tapers into the sharp cutting edge. The blade measures 8" (203 mm) (L) x ¾" (20 mm) (W). The overall length is 13" (330 mm) and it is made from standard grade stainless steel.
OAL: 13"
Catalog No. 50-193-1584 Supplier Electron Microscopy Sciences Supplier No. 7205903
This extra thick, rigid stainless steel blade is perfect for cutting hard tissue. Features a thick tapered edge with an optional 40 mm imprinted metric ruler. The top edge is 0.062" (1.6 mm) thick and tapers into the sharp cutting edge. The blade measures 8" (203 mm) (L) x ¾" (20 mm) (W). The overall length is 13" (330 mm) and it is made from standard grade stainless steel.
OAL: 13"