Learn More
Electron Microscopy Sciences EMS Heavy Pattern Dissecting Knife without Rule, 13" OAL
SDP
Catalog No. 501931583
Encompass_Preferred
This extra thick, rigid stainless steel blade is perfect for cutting hard tissue. Features a thick tapered edge with an optional 40 mm imprinted metric ruler. The top edge is 0.062" (1.6 mm) thick and tapers into the sharp cutting edge. The blade measures 8" (203 mm) (L) x ¾" (20 mm) (W). The overall length is 13" (330 mm) and it is made from standard grade stainless steel.
OAL: 13"
Catalog No. 50-193-1583 Supplier Electron Microscopy Sciences Supplier No. 7205902
This extra thick, rigid stainless steel blade is perfect for cutting hard tissue. Features a thick tapered edge with an optional 40 mm imprinted metric ruler. The top edge is 0.062" (1.6 mm) thick and tapers into the sharp cutting edge. The blade measures 8" (203 mm) (L) x ¾" (20 mm) (W). The overall length is 13" (330 mm) and it is made from standard grade stainless steel.
OAL: 13"