Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex HWL Foam Earplug Dispenser
SDP

Catalog No. 01259208
Change view
Click to view available options
Mounting:
Keyhole Slots
Magnetic
Wall Mountable
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
01-259-208 Wall Mountable
01-259-170 Magnetic
01-259-205 Keyhole Slots
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
N/A
Catalog No. 01-259-208 Supplier Poltex Supplier No. HWLMXEART
Only null left
Add to Cart
Add to Cart
N/A

Holds a box of foam earplugs and fits Honeywell Maximum Earplug S-7902 or equivalent. Choose from Wall Mount, HB Tape Mount, or Magnet Mount with durable PETG construction.

  • Holds a box of foam earplugs.
  • Fits Honeywell Maximum Earplug S-7902 or equivalent.
  • Choose from Wall Mount (screws/anchors included), HB Tape Mount (high-bond tape included), or Magnet Mount (powerful magnets included).
  • Durable and cleanable PETG construction.
  • Consumables not included.

Specifications

Color Clear
Dimensions (L x W x H) 5.5 x 8.4 x 8.25 in.
For Use With (Application) Holds A Box of Foam Earplugs
High Visibility No
Holds Box of Foam Earplugs
Quantity 1 Ea.
Weight (English) 1 lb.
Disposable Reusable
Metal Detectable No
Material PETG
Mounting Wall Mountable
Foldable No
Includes High Bond Tape for Mounting
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.