Learn More
Promocell GmbH PromoCell HWP-c 500,000 Human White Preadipocytes, visceral, cryopreserved
Supplier:  Promocell GmbH C12732
Encompass
Primary Human White Preadipocytes (HWP) are isolated from adult subcutaneous or visceral adipose tissue from different locations.Adipose tissue is crucial in energy storage and metabolic homeostasis. An increase in adipose tissue results either from an enlargement of mature adipocytes or from the differentiation of adipocyte precursor cells (preadipocytes) into new mature adipocytes. These preadipocytes are already present in adipose tissues. Since this differentiation process is controlled by a variety of growth factors and hormones, preadipocytes are suitable for the investigation of the physiological mechanisms controlling proliferation, differentiation, and function of adipose tissue.The PromoCell adipocyte cell culture system provides preadipocytes with optimized growth media and serum-free differentiation media.Our subcutaneous HWP are now also available from Diabetes Type I and Type II donors.  Our HWP are now also available from HLA-typed donors.
Catalog No. 50-306-062