Learn More
ProSci HWP1 Peptide

Description
Specifications
Specifications
| Gene ID (Entrez) | 3645386 |
| Accession Number | XP_712998.1 |
| Common Name | hyphal-specific cell wall protein; aka ECE2; C terminus similar to that of HWP99 and RBT1; similar to S. cerevisiae MUC1 (YIR019C) cell surface flocculin |
| Content And Storage | Store at -20°C. When reconstituted minimize freezing and thawing. |
| Format | Lyophilized |
| Formulation | HWP1 peptide is supplied as 100 μg of dried peptide. When the peptide is reconstituted in 200 μL water, the concentration will be 0.5 mg/mL |
| For Use With (Application) | Blocking Assay |
| Gene Symbol | HWP1 |
| Quantity | 0.1 mg |
| Sequence | HWP1 peptide corresponds to 15 amino acids of HWP1. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.