Learn More
Medchemexpress LLC HWY-289 | 908001-51-4 | 98.0% | 562.49 | 5 MG
SDP
Catalog No. 5000392516
Encompass_Preferred
HWY-289 is a semisynthetic protoberberine derivative that exhibits broad-spectrum and potent activities against phytopathogenic fungi, especially *Botrytis cinerea*. It works by altering the morphology of the mycelium and the internal structure of fungal cells, and it also reduces ATP content, ATPase activities, and key enzyme activities within the TCA cycle.
  • Exhibits broad-spectrum and potent activity against phytopathogenic fungi.
  • Particularly effective against *Botrytis cinerea*.
  • Alters the morphology of the mycelium and internal structure of fungal cells.
  • Reduces ATP content, ATPase activities, and key enzyme activities in the TCA cycle.
Catalog No. 50-003-92516 Supplier Medchemexpress LLC Supplier No. HY1634595MG
HWY-289 is a semisynthetic protoberberine derivative that exhibits broad-spectrum and potent activities against phytopathogenic fungi, especially *Botrytis cinerea*. It works by altering the morphology of the mycelium and the internal structure of fungal cells, and it also reduces ATP content, ATPase activities, and key enzyme activities within the TCA cycle.
  • Exhibits broad-spectrum and potent activity against phytopathogenic fungi.
  • Particularly effective against *Botrytis cinerea*.
  • Alters the morphology of the mycelium and internal structure of fungal cells.
  • Reduces ATP content, ATPase activities, and key enzyme activities in the TCA cycle.