Learn More
Medchemexpress LLC HWY-289 | 908001-51-4 | 98.0% | 562.49 | 50 MG
SDP
Catalog No. 5000392997
Encompass_Preferred
HWY-289 is a semisynthetic protoberberine derivative that exhibits broad-spectrum and potent activities against phytopathogenic fungi, particularly Botrytis cinerea (EC50 = 1.34 μg/mL). It alters the morphology of the mycelium and the internal structure of fungal cells. Additionally, it reduces ATP content, ATPase activities, and key enzyme activities in the TCA cycle.
  • Semisynthetic protoberberine derivative
  • Exhibits broad-spectrum antifungal activities
  • Potent against phytopathogenic fungi, especially Botrytis cinerea
  • Alters morphology of mycelium
  • Changes internal structure of fungal cells
  • Reduces ATP content
  • Reduces ATPase activities
  • Reduces key enzyme activities in the TCA cycle
Catalog No. 50-003-92997 Supplier Medchemexpress LLC Supplier No. HY16345950MG
HWY-289 is a semisynthetic protoberberine derivative that exhibits broad-spectrum and potent activities against phytopathogenic fungi, particularly Botrytis cinerea (EC50 = 1.34 μg/mL). It alters the morphology of the mycelium and the internal structure of fungal cells. Additionally, it reduces ATP content, ATPase activities, and key enzyme activities in the TCA cycle.
  • Semisynthetic protoberberine derivative
  • Exhibits broad-spectrum antifungal activities
  • Potent against phytopathogenic fungi, especially Botrytis cinerea
  • Alters morphology of mycelium
  • Changes internal structure of fungal cells
  • Reduces ATP content
  • Reduces ATPase activities
  • Reduces key enzyme activities in the TCA cycle