Learn More
TARGETMOL CHEMICALS INC HX 531 10MG
SDP
Catalog No. 502577176
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. HX 531 is a potent antagonist of the retinoid X receptor with an IC50 value of 18 nM. purity: 99%
Catalog No. 50-257-7176 Supplier TARGETMOL CHEMICALS INC Supplier No. T2284310MG
Also available in 1 mg 5 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. HX 531 is a potent antagonist of the retinoid X receptor with an IC50 value of 18 nM. purity: 99%