Learn More
Apexbio Technology LLC HX 531 188844-34-0 10mg
SDP
Catalog No. 5000088931
Encompass_Preferred
HX 531 (CAS 188844-34-0) is a small-molecule antagonist targeting the retinoid X receptor (RXR) It modulates RXR-associated signaling pathways thereby influencing both tumorigenic and metabolic biological processes HX 531 exerts its biological activity by antagonizing RXR which results in cell cycle arrest at the G0/G1 phase and modulation of leptin-dependent pathways and the p53-p21Cip1 signaling axis In rodent models HX 531 demonstrates an inhibitory activity with an IC50 value of approximately 18 nM Based on these pharmacological properties HX 531 holds research potential in the study of metabolic disorders obesity diabetes and cancer biology
Catalog No. 50-000-88931 Supplier Apexbio Technology LLC Supplier No. B549410
HX 531 (CAS 188844-34-0) is a small-molecule antagonist targeting the retinoid X receptor (RXR) It modulates RXR-associated signaling pathways thereby influencing both tumorigenic and metabolic biological processes HX 531 exerts its biological activity by antagonizing RXR which results in cell cycle arrest at the G0/G1 phase and modulation of leptin-dependent pathways and the p53-p21Cip1 signaling axis In rodent models HX 531 demonstrates an inhibitory activity with an IC50 value of approximately 18 nM Based on these pharmacological properties HX 531 holds research potential in the study of metabolic disorders obesity diabetes and cancer biology